Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Ladders and Step Stools

https://ca.vwr.com/stibo/smallweb/std.lang.all/53/24/18945324.jpg

Safely access hard-to-reach items using the ladders and step stools offered in various height capacities. The overall lightweight construction provides optimum portability between sites. Multi-position ladders feature heavy gusset bracing to securely lock desired height. The rubber platform step on stools prevents unwanted movement when in use for stable retrieval. Non-skid floor rings or retracting casters models assist in keeping stool secure when weight is applied. The ladders and step stools are essential in any application environment.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"9"
  List View Easy View
Sort by:
 
 
 
 

Catalog Number: (59075-022)

Supplier:  Bel-Art Products
Description:   This handy step stool provides safe, easy access to high shelves, drawers, equipment and other hard-to-reach areas.
Catalog Number: (470206-038)

Supplier:  Cramer
Description:   As soon as weight is applied, the easy-moving casters retract for firm and stable support.
Supplier:  Cramer
Description:   These rolling step stools are constructed of solid steel with a 226.8kg (500lb.) downloading rating
Small Business Enterprise
Supplier:  Lakeside
Description:   1" chrome plated steel construction is durable and easy to clean
Catalog Number: (470094-486)

Supplier:  BOSTWICK-BRAUN COMPANY
Description:   These step stool won’t rust or dent. Bottom pads help prevent stool from slipping and protect floors. Holds up to 300 lbs. Easy to store in cupboard o...
Catalog Number: (59075-005)

Supplier:  Cramer
Description:   Wrap-around bumper for use with Cramer Kik-Step step stools. Does not require adhesive.
Small Business Enterprise
Supplier:  Ballymore
Description:   A safe and convenient fixture for any laboratory
Catalog Number: (24450-004)

Supplier:  Bandy
Description:   Lightweight, two-level step stool for the cleanroom
Product available on GSA Advantage® Minority or Woman-Owned Business Enterprise Small Business Enterprise
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 9
  1  2  Next 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping