Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "mortars"

https://ca.vwr.com/stibo/smallweb/std.lang.all/95/71/18989571.jpg

Mortars are valuable grinding vessels used since ancient times in preparation of crushed and ground samples for experimentation. These can come in a variety of forms, including those used by hand, in conjunction with a machine, or after being frozen with liquid nitrogen for use in procedures dealing with frozen tissues. Mortars, and particularly mortar grinders, are ideal for grinding dry samples and solid materials in suspension, and for manufacturing and homogenizing pastes and creams to laboratory standards.

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"3"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Bel-Art Products
Description:   Grind frozen samples without liquid nitrogen hazards.
Supplier:  SILICO & CHEMICO PORCELAIN SE
Description:   MORTAR PORCELAIN 70 MM 50ML.
Catalog Number: (14213-042)
Supplier:  Retsch
Description:   Designed for use with Retsch® RM200 Mortar Grinder, this hardened steel mortar is intended for use in less demanding applications, or under rough cond...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 3  of 3