Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • GSA Contract (3) SBE (4) SBE (1) UL Listed (5)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Pipet Filler Bulbs

https://ca.vwr.com/stibo/smallweb/std.lang.all/96/18/18709618.jpg

Pipette filler bulbs have replaced unsafe mouth pipetting, by introducing a manual liquid aspiration transfer using simple squeezing and releasing action. The brightly colored bulbs are typically constructed out of natural rubber compatible with volatile laboratory fluids. Models featuring arrows on the blowout valves eliminate confusion when operating. Able to operate with one hand, the versatile dispensing tools work well with a wide range of style and size pipettes.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"24"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   Made of durable natural rubber, bulbs are suitable for medicine droppers and small dropping pipets.
Small Business Enterprise
Catalog Number: (82024-574)
Supplier:  VWR International
Description:   Designed for use with small and large pipets, bulb features a white plastic attachment with a silicone conical adapter for safe and secure attachment ...
Supplier:  VWR International
Description:   Small-volume, natural rubber latex bulbs are designed for use with Pasteur pipets, small pipets, and medicine droppers.
Supplier:  VWR International
Description:   Pipet fillers do away with the frequently hazardous practice of drawing liquids into pipets by mouth.
Product available on GSA Advantage®
Catalog Number: (53497-055)
Supplier:  VWR International
Description:   Pipet filler for all pipets from 0.05 to 100 mL.
Catalog Number: (53502-205)
Supplier:  VWR International
Description:   Pipet fillers do away with the frequently hazardous practice of drawing liquids into pipets by mouth.
Product available on GSA Advantage®
Supplier:  VWR International
Description:   Easy-to-grip bulbs feature heavy-walled necks for use with pipets and syringes.
Catalog Number: (53502-200)
Supplier:  VWR International
Description:   This polypropylene pipet bulb is acid- and alkali-resistant, always ready to use, and requires little maintenance
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 24
  1  2  3  Next