Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • SBE (2)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Bottle Racks

https://ca.vwr.com/stibo/smallweb/std.lang.all/49/70/18944970.jpg

Bottle racks organize and securely hold contained liquids for immediate access storage or easy transportation. The lightweight and durable construction is also resistant for even the most corrosive chemicals. Different shaped styles may be conveniently stored on benchtops or mounted on walls to save space. Extended handles allow carrier racks to relocate bottles between working site safely. The bottle racks hold different quantities of bottles in different sizes.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"14"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   Maximize bench space with the Quad Bottle Organizer.
Supplier:  VWR International
Description:   Tough, lightweight carrier holds up to four wash bottles.
Supplier:  VWR International
Description:   Utilize vertical space of your benchtop with the Workstation Riser.
Supplier:  Bel-Art Products
Description:   Small enough to hold in one hand and enough choices to accommodate most vials and small bottles.
Supplier:  Bel-Art Products
Description:   This clear acrylic rack organizes valuable USP™ reference standards while allowing easy viewing of bottle labels.
Supplier:  Bel-Art Products
Description:   Racks have 12 places (3x4 rows) for 300 mL Biological Oxygen Demand bottles.
Supplier:  Bel-Art Products
Description:   This organizing rack system is a highly valuable aid for refrigerated storage of small bottles of blood-typing sera, chemistry controls, enzyme reagen...
Catalog Number: (89174-476)
Supplier:  DWK Life Sciences
Description:   Manufactured from rugged and durable PVC coated wire, this rack is designed to accommodate 300mL BOD bottles
Small Business Enterprise
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 14
  1  2  Next