Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Centrifuge Tube Racks

https://ca.vwr.com/stibo/smallweb/std.lang.all/95/25/18709525.jpg

Safely hold samples before and after separating spinning cycles with the centrifuge tube racks. Organize the conical bottom tubes in different placement matrixes or wire styles offered. While some are designed for a specific common size, other centrifuge tube racks have the capability of accommodating multiple different diameter size tubes for testing flexibility. The autoclavable tools feature inventory labels and different colored manufacturing for correct identification after transportation.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"9"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   These racks are designed to hold VWR® centrifuge tubes and replace foam racks
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Product is manufactured with low CO₂ impact Product available on GSA Advantage®
Catalog Number: (89199-176)
Supplier:  VWR International
Description:   The VWR® Dual Centrifuge Tube Stand is capable of holding both 15mL and 50mL centifuge tubes.
Supplier:  VWR International
Description:   Polypropylene racks are available in seven colors, and are also offered in an assorted color pack with one of each color: blue, green, orange, red, an...
Supplier:  VWR International
Description:   Durable stainless steel freezer racks facilitate organization and maximize valuable storage space.
Small Business Enterprise
Supplier:  Bel-Art Products
Description:   The only wire rack that holds 500 mL conical centrifuge tubes.
Supplier:  Globe Scientific
Description:   Rack is designed to hold both 15 and 50mL centrifuge tubes simultaneously
Catalog Number: (76040-426)
Supplier:  Argos Technologies
Description:   Ideal for use in 3" boxes or directly on the benchtop.
Supplier:  Simport Scientific
Description:   The UniRack™ can hold up to 25 polystyrene or polyprolylene 10 and 12 mm tubes
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 9
  1  2  Next