Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "cuvette-racks"

https://ca.vwr.com/stibo/smallweb/std.lang.all/50/94/18945094.jpg

Scratch-proof manufactured cuvette racks store fragile and expensive laboratory equipment securely. With minimized contact and steam autoclaving ability, the horizontal or sloped designed racks reduce the risk of damage to samples. The supports offer immediate access to required spectroscopic assaying devices when experimenting speed is paramount. Although non-stacking options are available, stackable styles save precious benchtop working space. Stands accommodate different quantities of any sized cuvette in various matrixes and number of places.

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"5"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   These two-tier racks support cuvettes with a minimum of physical contact, greatly reducing the chance of damage.
Catalog Number: (89000-626)
Supplier:  VWR International
Description:   This polyethylene rack accommodates twelve 10 mm cuvettes, and also holds twelve cuvettes horizontally for storage and protection.
Supplier:  Bel-Art Products
Description:   This polypropylene rack holds twenty 10 mm light path spectrophotometer cuvettes in two tiers with numbered wells.
Supplier:  Bel-Art Products
Description:   All-polypropylene rack provides gentle no-scratch support for fragile and expensive spectrophotometer cuvettes.
Catalog Number: (CA97024-722)
Supplier:  PerkinElmer
Description:   Cuvette Rack^
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 5  of 5