Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • Environmentally Preferable (4) GSA Contract (2) UL Listed (4)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Test Tube Racks

https://ca.vwr.com/stibo/smallweb/std.lang.all/53/54/18945354.jpg

Compatible with automated equipment, test tube racks can be loaded with maximum efficiency. The single piece options provide constant sample visibility in coated wire or transparent plastic styles. With heat resistance, the test tube racks can even hold the glassware during autoclaving procedures. The various diameter sized placement holes are separated so that the tube edges will not overlap and may feature an alphanumerical label grid for the best sample identification.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"124"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   No assembly required - racks are a single piece molded from either nylon 6 glass fiber reinforced material or polypropylene.
Supplier:  VWR International
Description:   A great alternative to wire racks, with price- and space-saving advantages over other plastic racks.
Supplier:  VWR International
Description:   Chemical and corrosion-resistant steel wire racks are coated with heat-cured epoxy.
Catalog Number: (82024-452)
Supplier:  VWR International
Description:   Rack is ideal for holding up to eight 50 mL conical centrifuge tubes and features a slotted bottom for secure handling.
Supplier:  VWR International
Description:   These polypropylene (PP) plastic racks provide a practical method of pipetting and storing a large number of samples.
Supplier:  VWR International
Description:   This black epoxy-coated rack of heavy-gauge steel wire holds 30 mm diameter or smaller centrifuge tubes in a 4 x 6 pattern, 24 tubes per rack.
Catalog Number: (82024-496)
Supplier:  VWR International
Description:   Ideal for bench work or storage, these polypropylene racks are reinforced with fiberglass to make them strong and versatile.
Supplier:  VWR International
Description:   VWR® Aluminum Test Tube Racks are punched from hard sheet aluminum.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 124
  1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  Next