Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Cryogenic Straws

https://ca.vwr.com/stibo/smallweb/std.lang.all/68/70/20236870.jpg

Inert to extreme temperatures, cryogenic canes are devices that accommodate common sized vials for preservative storage. Snap cryovials into place for a reliable seal that will remain hermetic even in liquid nitrogen chilling. Available in different working lengths to fit any freezer, the color coded cryogenic canes assist with the proper identification of hazardous or biological material samples. Achieve additional security when handling by adding the transparent sleeves that support fast empty space location.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"5"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  Thermo Scientific
Description:   Firmly holds NALGENE® cryogenic vials for storage in Dewar-type, liquid-nitrogen freezers.
Supplier:  Thermo Scientific
Description:   Clear plastic sleeve encloses a NALGENE® CryoCane™ for extra security during handling and storage

Supplier:  Thermo Scientific
Description:   Flexible polyethylene tubing provides extra safety when freezing hazardous samples or storing in liquid nitrogen
Catalog Number: (CA24270-015)

Supplier:  Thermo Scientific
Description:   Made of aluminum
Supplier:  Simport Scientific
Description:   For storage of up to five 1.2 or 2 mL Simport Cryovial® tubes in liquid nitrogen containers such as Dewar flasks.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 5  of 5
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping