Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "labeling-tapes"

https://ca-bot.vwr.com/stibo/smallweb/std.lang.all/94/60/18989460.jpg

Labeling tapes adhere to any dry surface and are compatible with most writing instruments. Resistant to harsh liquids, temperatures, or testing environments, the adhesives guarantee that samples will be correctly identified regardless of the procedure involved. Essential for routine use, the pressure-sensitive labeling tapes tear easily at any desired size and come in different color options for simple color coding. Standard roll sizes accommodate use with dispensers for maximum convenience.

SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
 
 
SearchResultCount:"35"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   VWR® General-Purpose Laboratory Labeling Tape sticks to virtually any surface, can be marked with most writing utensils, and reliably withstands extre...
Supplier:  VWR International
Description:   These 9 mm x5m, vinyl, self-stick tapes color code autoclave items.
Catalog Number: (10159-644)
Supplier:  VWR International
Description:   VWR® Preprinted Flammable Warning Tape is imprinted with red ink on white tape and can be used on virtually any surface where visual warning is requir...
Supplier:  Bel-Art Products
Description:   Adheres to any clean surface, including Teflon® fluoropolymer resin-coated materials, and peels off without leaving a sticky residue.
Supplier:  BURKLE INC
Description:   CLOSE-IT TAPE 150MM
Supplier:  Sklar
Description:   I.D. Sheet Tape is latex-free and nonsterile.
Supplier:  Sklar
Description:   I.D. Roll Tape is latex-Free and nonsterile.
Supplier:  Brady Worldwide
Description:   Two Color Print Ribbon Cartridges - 200 ft/roll. Brady has super-tough smear-proof ribbons assure your printed text lasts for years outdoors and in an...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 35
  1  2  3  4  5  Next