Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Product Category
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "test-papers"

https://ca.vwr.com/stibo/smallweb/std.lang.all/94/31/18989431.jpg

By the simple appearance of color, test papers provide fast conclusions on sample component levels. Designed for specific element or multi-parameter detection, the reagent- or indicator-treated reaction papers give a general content range using graduated intervals. Dip the disposable sticks into the given sample, then match the color to the provided chart for the most economical and easiest method in quality testing. Having an overall compact size, the test papers can be used on-site.

SearchPresentationType-SearchPresentationType-SearchPresentationType-VERTICAL
 
 
SearchResultCount:"83"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  BDH
Description:   Used for rapid pH testing directly at the point of interest, strips are always ready for immediate use.
Supplier:  BDH
Description:   These indicator papers show two different colors for each pH value at intervals of 0.3–1 pH unit, increasing the accuracy of readings and allowing goo...
Catalog Number: (CA71009-070)
Supplier:  VWR
Description:   Accutint™ pH test paper is supplied in a poly dispenser containing a continuous 4.6 meter strip with an individual colour chart on the label. Test pap...
SDS
Supplier:  VWR
Description:   No Color Chart Necessary.
Supplier:  Avantor Performance Materials
Description:   J.T.Baker® brand pH indicator sticks feature up to four color-fixed test zones on a durable plastic strip.
Supplier:  Avantor Performance Materials
Description:   BAKER-pHIX pH papers feature convenient indicators with a color scale printed on the paper for quick accurate readings.
Supplier:  Avantor Performance Materials
Description:   J.T.Baker® brand DUAL-TINT pH papers offer easy-to-read, precise pH indication in a rolled 17-foot strip, 3⁄8” wide in a refillable dispenser with col...
Supplier:  MilliporeSigma
Description:   These strips provide a convenient method for accurate colorimetric pH indication.
SDS
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 83
  1  2  3  4  5  6  7  8  9  10  11  Next