Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • GSA Contract (11)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "transfer-membranes"

https://ca.vwr.com/stibo/smallweb/std.lang.all/99/35/18989935.jpg

Highly sensitive transfer membranes use capillary action methods to separate DNA, RNA, or protein fragments for detection. Working well with the gels and buffers required for transfer and immobilization steps, the transfer membranes are durable enough to be subjected to multiple stripping, reprobing, hybridization cycles. The high-binding discs, sheets, and rolls come in various pore sizes and charges to fit any blotting techniques in molecular biology and genetic research.

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"32"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR
Description:   Totalblot Pvdf 1Roll^
SDS
Supplier:  Pall Laboratory
Description:   Membranes have a high binding affinity (209µg/cm2) and are ideal for Western blot confirmatory tests.
SDS
Supplier:  Pall Laboratory
Description:   Provide high sensitivity and low background for enhanced detection and resolution.
Supplier:  Azure Biosystems
Description:   Azure offers pre-cut PVDF and nitrocellulose membranes for Western blotting applications.
Product available on GSA Advantage®
Supplier:  Pall Laboratory
Description:   Pure unsupported nitrocellulose membrane ideally suited for colony and plaque lifts and protein transfers.
SDS
Supplier:  GE Healthcare - Life Sciences
Description:   Amersham Hybond P 0.45 PVDF is a 0.45 μm pore size hydrophobic membrane, with high physical strength, highly suitable for stripping and reprobing. Mem...
SDS
Supplier:  GE Healthcare - Life Sciences
Description:   Amersham Protran membranes are manufactured from 100% pure nitrocellulose to ensure the highest binding capacity possible. They are compatible with a ...
SDS
Supplier:  GE Healthcare - Life Sciences
Description:   Amersham Hybond P 0.2 PVDF is a 0.2 μm pore size hydrophobic membrane with high physical strength. This results in significant handling advantages ove...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 32
  1  2  3  4  Next