Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Fermentation Tubes

https://ca.vwr.com/stibo/smallweb/std.lang.all/93/54/18989354.jpg

Fermentation tubes are used to verify gas production in fermentation exercises, with gas produced by yeast or other organisms rising into the evolution tube, and they are available in graduated or ungraduated formats, with or without a foot. Fermentation tubes are manufactured from borosilicate glass that is durable and fully autoclavable for easy sterilization. Certain models are also available with additional orbs in their design and extensions.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"4"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  DWK Life Sciences (KIMBLE®)
Description:   KIMAX® fermentation tubes are used to collect gas in a broth culture formed by microorganisms with a bulb, a glass foot, and vertical tube that has a ...
Catalog Number: (470104-226)

Supplier:  G.S.C. INTERNATIONAL, INC.
Description:   TUBE FERMENTATION 100X13MM UNGRAD W/FOOT
Supplier:  G.S.C. INTERNATIONAL, INC.
Description:   Verify gas production in fermentation exercises, gas produced by yeast or other organisms rises into a 5 or 10 mL evolution tube.
Catalog Number: (470235-722)

Supplier:  G.S.C. INTERNATIONAL, INC.
Description:   Fermentation tubes manufactured from borosilicate glass.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 4  of 4
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping