Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "melting-point-capillaries"

https://ca.vwr.com/stibo/smallweb/std.lang.all/95/12/18989512.jpg

Melting point capillaries are sealed with samples inside and placed inside a melting point apparatus, which is then heated in order for the user to observe any phase changes from solid to liquid and when they occur. Tubes are manufactured from borosilicate glass and feature openings on one or both ends. These are quality products perfect for the handling and transport of small volumes of liquid, melting point determinations, and many other purposes. The tubes are uniform and are cut at right angles.

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"6"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Wilmad-LabGlass
Description:   Fabricated from low extractable borosilicate glass, these melting point capillary tubes will not react with organic compounds. Capillary tubes are clo...
Supplier:  Wilmad-LabGlass
Description:   A modified version of the Thiele tube, this model improves and increases the rate of circulation of the bath liquid during melting point determination...
Supplier:  Wilmad-LabGlass
Description:   Used for melting point determination tests, this borosilicate glass Thiele tube resembles a glass test tube with an attached handle.
Supplier:  CORNING CA
Description:   Quality Glassware at an Affordable Price.
Product available on GSA Advantage®
Supplier:  Chemglass
Description:   Tubes are manufactured from borosilicate glass and feature openings on both ends. Tubes are packaged 100 per vial. The 0.25mm thick tube conforms to U...
Small Business Enterprise
Supplier:  Buchi
Description:   These precision glass capillaries ensure highly reproducible melting point determinations.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 6  of 6