Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • GSA Contract (23) MWBE (5) SBE (181) SBE (955)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Chromatography Vials

https://ca.vwr.com/stibo/smallweb/std.lang.all/40/36/18374036.jpg

Designed to use separating organic and inorganic compounds for analysis, chromatography vials come in different finishes to guarantee chemical compatibility. Autosampler compatible closures come in different colors, sizes, and septa materials. Offered with a write-on patch, possible identification errors are eliminated. Supporting use with inserts or already possessing a conical interior shape, maximum retrieval of contents to eliminate waste. Select from white, amber, or clear for trouble-free mixture and solvent observation.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"128"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   Vials are specifically designed with a crimp cap profile for robotic arm autosamplers but have easy-to-use screw-thread caps.
Supplier:  VWR International
Description:   Standard opening crimp-top vials have a maximum fill volume of 1.8 mL and are manufactured from inert, 33 expansion borosilicate glass.
Supplier:  VWR International
Description:   Wide opening vials feature 40% larger neck opening than standard opening vials, for easier access and reduced needle damage.
Supplier:  VWR International
Description:   Standard opening screw-thread vials are manufactured from borosilicate glass and have a maximum fill volume of 1.8 mL.
Supplier:  VWR International
Description:   Vials feature a versatile neck that allows closure by either snap-cap seals or aluminum crimp seals.
Catalog Number: (66015-702)
Supplier:  VWR International
Description:   Designed for use in Alcott or Waters® autosamplers, the clear snap-plug closures of these shell vials can be pierced with standard autosampler syringe...
Supplier:  VWR International
Description:   Designed for use with Waters® WISP® (48-position) and Shimadzu™ autosamplers.
Supplier:  VWR International
Description:   These headspace vials are suitable for use with all common autosamplers.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 128
  1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  Next