Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "crimpers"

https://ca.vwr.com/stibo/smallweb/std.lang.all/40/38/18374038.jpg

Stainless steel crimpers are designed to manually secure standard crimp closures on vials. This easy to use tool features cushioned handles to offer maximum comfort. Many models can adjust the sealing compression. Autoclavable, tapered jaws, and easy identification labeling styles are available. Achieve seals with minimum work with our electric powered crimpers that are also available.

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"18"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  DWK Life Sciences
Description:   Crimpers feature polished crimping jaws for consistent sealing, a redesigned spring assembly, and a short, cushioned ergonomic handle that helps reduc...
Small Business Enterprise
Supplier:  Thermo Scientific
Description:   Tools are designed to manually secure or release standard crimp seals to and from vials. Crimper is adjustable to accommodate various septa thicknesse...
Supplier:  DWK Life Sciences
Description:   These crimpers are used for flip cap seals and for aluminum seals sizes 16.5, 28.5, and 30 mm.
Small Business Enterprise
Supplier:  Thermo Scientific
Description:   Attach or remove aluminum seals with the single press of a button. Electronic crimpers and decrimpers offer adjustable crimp settings compatible with ...
Supplier:  Ace Glass
Description:   This decapper features a new ergonomic design cushioned handle that aids in reducing hand fatigue and provides a higher degree of comfort for the user...
Product available on GSA Advantage® Small Business Enterprise
Supplier:  Thermo Scientific
Description:   Achieve secure, reproducible crimps with the push of a button by using programmable electronic high-power crimp stations. A reliable crimp is guarante...
Supplier:  Thermo Scientific
Description:   Achieve secure, repeatable crimps or remove seals with minimized risk of damage by using stainless steel manual crimpers and decrimpers. crimpers prov...
Supplier:  MicroSolv Technology Corp
Description:   These manual crimping tools are easy to use. Use with 11mm or 20mm aluminum crimp caps to achieve great seals for autosampler vials. The crimping jaws...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 18
  1  2  3  Next