Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Capillary Viscometers
https://ca.vwr.com/stibo/smallweb/std.lang.all/49/90/18944990.jpg
Calibrated with clear markings, the capillary viscometers support visual viscosity measurements of liquids. The direct or reverse flow styles have respectively positioned reservoirs to calculate the time it takes a specific fluid quantity to flow through the capillary. Since the driving force is gravity and cannot be varied, the standardized glass tubes produce more reliable results. Common Ostwald and Ubbelohde capillary viscometers are inexpensive alternatives to digital versions when determining material properties.
SearchPresentationType-VERTICAL
SearchResultCount:"20"
  List View Easy View
Sort by:
Catalog Number: (66044-007)
Supplier:  VWR International
Description:   For determination of relative viscosities by measuring time of flow of a definite volume of fluid through a standard capillary.
Supplier:  Ace Glass
Description:   For use in obtaining kinematic viscosities
Product available on GSA Advantage® Small Business Enterprise
Supplier:  DWK Life Sciences (KIMBLE®)
Description:   For obtaining kinematic viscosities of transparent liquids (ASTM Method of Test D445)
Supplier:  Ace Glass
Description:   Used in ASTM Method D 1601
Product available on GSA Advantage® Small Business Enterprise
Supplier:  Wilmad-LabGlass
Description:   Quickly and easily measure viscosities of Newtonian liquids with these borosilicate glass capillary tube viscometers.
Supplier:  DWK Life Sciences (KIMBLE®)
Description:   For rough control work where bubble comparison is made to limit varnish samples of known viscosity, reference ASTM D 1545
Supplier:  Fungilab
Description:   Cannon-Fenske routine viscometers quickly and easily measure the viscosities of transparent Newtonian liquids in the chemical and petroleum industries...
Supplier:  Technical Glass Products
Description:   These reverse flow type viscometers permit measurement of dark liquids whose meniscus cannot be seen at the timing marks when using a Cannon-Fenske ro...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 20
  1  2  3  Next 