Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Wall Hooks

https://ca.vwr.com/stibo/smallweb/std.lang.all/00/07/18990007.jpg

Convenient wall hooks hang apparel or equipment to keep accessible at all times. Maintain tidiness and general organization of storage areas with garment racks featuring hooks for one to four items. Choose decorative options of brushed metal or wood, or select functional products of heavy duty molded plastic or stainless steel. Based on the wall surface, mounting is available in adhesive backing, magnetic options, or keyhole mounts for compatibility.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"7"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   Cylinder wall brackets secure 10.2 – 30.5 cm (4 – 12") diameter cylinders using 137.2L x 3.8Wcm (54 x 11/2") polypropylene strap...
Supplier:  VWR International
Description:   Cylinder wall-mount brackets are constructed of cast aluminum and contoured to allow cylinders to fit firmly along the support edge.
Product available on GSA Advantage®
Supplier:  Bel-Art Products
Description:   This hook is perfect for hanging lab coats, aprons, or other clothing.
Supplier:  TrippNT
Description:   Single hook. Magnetic back. Ideal for holding lab coats, masks, tubing, and more. Made from solid PVC plastic. Hooks can be installed instantly through the use of the magnet backing. Dimensions: 10.2Lx16.5Dx5.8Hcm. Load capacity: 4.5kg.
Minority or Woman-Owned Business Enterprise Small Business Enterprise
Catalog Number: (RK21984)
Supplier:  Restek
Description:   Optional wall mounting bracket allows for the mounting of Super Clean® base plates with the use of screws.
Supplier:  Justrite
Description:   Ideal for applications utilizing safety disposal cans with disconnects.
Supplier:  QUANTUM STORAGE SYSTEMS
Description:   Accepts all sizes of Stack and Hang bins and Mesh bins.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 7  of 7