Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Blotting Apparatus

https://ca.vwr.com/stibo/smallweb/std.lang.all/49/56/18944956.jpg

VWR offers a variety of different blotting apparatus for Southern, Western, and Northern dot blotting procedures. Electroblotting systems feature easy blotting of multiple gels simultaneously in less than an hour. Mini and large tank systems produce uniform and consistent results. Semi-dry blotting systems offer fast and efficient transfers of proteins to membranes with low buffer volume. Most blotting apparatus include essential accessories.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"24"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   Mini electroblotting system permits quick and easy blotting of three gels simultaneously.
Catalog Number: (CA89129-238)
Supplier:  GE Healthcare - Life Sciences
Description:   PR 648 Slot Blot Filtration Manifolds are designed for ease of use and maximum filtration with minimum chance of cross contamination
Catalog Number: (CA89129-114)
Supplier:  GE Healthcare - Life Sciences
Description:   Including valves, connectors and tubing.
Supplier:  GE Healthcare - Life Sciences
Description:   TurboBlotter* Refill, membrane, large, Nytran* SuPerCharge 0. 45um, 15x15 cm, Reusable blotting device requires less buffer and blotting paper, Downward capillary transfer allows for alkaline buffer transfers in one hour and neutral transfers in 3 hour, Pack of 2
SDS
Supplier:  GE Healthcare - Life Sciences
Description:   Amersham ECL Semi-Dry Blotters are designed to deliver outstanding electrophoretic transfer results using low current and voltage with a minimal amoun...
Supplier:  GE Healthcare - Life Sciences
Description:   VacuGene XL Vacuum Blotting System is the choice for nucleic acid blotting.
Catalog Number: (CA95044-782L)
Supplier:  GE Healthcare - Life Sciences
Description:   To assure a uniform electrical field, Mini Tank Transfer Unit has 80 cm of wire in seven parallel strands on each electrode panel
Catalog Number: (CA95044-746L)
Supplier:  GE Healthcare - Life Sciences
Description:   Perform 1-D and 2-D electrophoresis and electrotransfer in a single compact unit.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 24
  1  2  3  Next