Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • GSA Contract (17) SBE (2) UL Listed (7) C-UL Listed (7) CSA Certified (7) Irregular Voltage (19)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Microcentrifuges

https://ca.vwr.com/stibo/smallweb/std.lang.all/40/57/18374057.jpg

Compact microcentrifuges isolate DNA or simply separate micro volume mixtures. Load microtubes and standard or dell-welled plates in the instruments easily. Adaptable to any industrial or laboratory setting, non-refrigerated or refrigerated models come with multiple mode options for the high-speed and powerful rotors. Continuous or pulse spinning options move samples fast and consistently while included timers alert with sample preparation is complete. Microcentrifuges may come with or without the protective lid.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"24"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   The VWR® Mini Centrifuge is designed for the centrifugation of microcentrifuge tubes and PCR strips.
Catalog Number: (75838-336)

Supplier:  VWR International
Description:   The compact, affordable VWR High Speed Microcentrifuge is capable of high speed performance up to 15,500 rpm.
Supplier:  VWR International
Description:   The VWR® mini centrifuge is designed for quick spin-downs of microcentrifuge tubes and PCR strips and tubes.
Supplier:  VWR International
Description:   These compact microcentrifuges are ideal for DNA, RNA, and temperature-sensitive samples.
Product available on GSA Advantage®
Catalog Number: (10144-214)

Supplier:  VWR International
Description:   Spins at 2500 rpm/ 850 xg, to quickly spin down droplets and condensation in PCR plates

Supplier:  Beckman Coulter
Description:   This non-refrigerated, cold-room compatible, high-performance microcentrifuge is ideal for fast pelleting or isolation of DNA, RNA, proteins, and viru...
Supplier:  HETTICH INSTRUMENTS
Description:   The Hettich Lab Technology MIKRO 220 microliter centrifuge series offers large capacities and high-performance for high-throughput laboratories.
Product available on GSA Advantage®
Supplier:  HETTICH INSTRUMENTS
Description:   The Hettich MIKRO 200 microliter centrifuge series is amongst the highest performing in its class.
Product available on GSA Advantage®
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 24
  1  2  3  Next 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping