Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Kjeldahl Distillation Systems

https://ca.vwr.com/stibo/smallweb/std.lang.all/94/54/18989454.jpg

Overcoming the original testing sensitivity issues, the Kjeldahl distillation systems conduct nitrogen analyses that determine the total protein content with unparalleled precision and reproducibility. Available in various levels of automation, the temperature-resistant equipment purifies diluted liquids through steam distilling. The compact benchtop units have customizable technology to meet specific testing requirements and industry regulations. Completing the final step in Kjeldahl method processing, the versatile distillation systems can be used in many fields.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"6"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VELP SCIENTIFIC INC.
Description:   UDK129 Distillation Unit 115V/50-60Hz^
Supplier:  VELP SCIENTIFIC INC.
Description:   UDK139 Distillation Unit 230V/50-60Hz^
Supplier:  VELP SCIENTIFIC INC.
Description:   UDK149 Distillation Unit 230V/50-60Hz^
Supplier:  Labconco
Description:   Designed for mounting on a base cabinet or table, unit provides durability for laboratories which require a separate distillation station.
Product available on GSA Advantage®
Supplier:  Labconco
Description:   Designed for mounting on a base cabinet or table, unit provides durability for laboratories which require a separate distillation station.
Product available on GSA Advantage®
Catalog Number: (76169-012)
Supplier:  Buchi
Description:   The standalone Kjeldahl BUCHI model K-355 is versatile to meet a variety of laboratory needs
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 6  of 6