  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Scrubber Systems
https://ca.vwr.com/stibo/smallweb/std.lang.all/97/75/18989775.jpg
Jet scrubber systems clean up any final vapor traces in reaction vessels after digestion and distillation procedures. Effectively washing and neutralizing any residual aggressive or toxic fumes, the scrubbers pump liquid into the laboratory equipment for safe reuse. The separate components are easy to clean and guarantee that exhaust and air cleaning tasks run smoothly. With a small footprint, the gas scrubber systems can fit any work space needed.
SearchPresentationType-VERTICAL
SearchResultCount:"1"
  List View Grid View Easy View (new)
Sort by:
Catalog Number: (76169-020)
Supplier:  Buchi
Description:   The K-415 TripleScrub is a three-step gas scrubbing instrument with a powerful pump and condensation, neutralization and adsorption steps
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 1  of 1