Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Oxygen Meters

https://ca.vwr.com/stibo/smallweb/std.lang.all/40/42/18374042.jpg

Automatic or manual dissolved oxygen meters measure the oxygen level present to evaluate the water quality. Carry these portable tools between sites use the simple interface guided navigation to capture multiple readings for concise comparisons. The waterproof housing and probe are ruggedly designed to operate one-handedly. Dissolved oxygen meters may feature additional low battery indicators and dual sensors to also record temperatures. The sizes, ranges, and resolutions may differ among supplied models.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"21"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalog Number: (21800-022)
Supplier:  VWR International
Description:   This dissolved oxygen meter simultaneously displays oxygen concentration and temperature.
Catalog Number: (21800-012)
Supplier:  VWR International
Description:   Two displays simultaneously show conductivity reading and temperature measurements on 34.9 mm (13/8") LCD readout.
Catalog Number: (CA10146-160)
Supplier:  Hach
Description:   Portable Meter Kit HQ40d with LDO10101 Optical Dissolved Oxygen Probe and PHC10101 pH Electrode, KTO, 1m Cables
Supplier:  YSI
Description:   EcoSense Optical Dissolved oxygen and Temperature Field Probe, 1M Cable Kit; ODissolved oxygen 200 Instrument, 1-meter Cable and Probe, and hard-sided Carrying Case, Grey
Supplier:  Mettler Toledo
Description:   <bold>This dissolved oxygen meter features linear and non-linear calibration, 100% and 0% calibration, and the ability to store 200 data sets
Supplier:  IN-SITU, INC.
Description:   The smarTROLL Rugged Dissolved Oxygen (RDO) Handheld combines optical dissolved oxygen (DO) technology with revolutionary smartphone mobility
Supplier:  Mettler Toledo
Description:   <bold>SevenGo® Pro™ Optical DO meter is based on optical RDO® (Rugged Dissolved Oxygen) technology, it makes the measurement easier
Supplier:  YSI
Description:   Microprocessor-based, menu-driven dissolved oxygen meters are designed for laboratory measurement of dissolved oxygen and are useful for B.O.D
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 21
  1  2  3  Next