Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Gas Detection Systems

https://ca.vwr.com/stibo/smallweb/std.lang.all/52/76/18945276.jpg

The pharmaceutical industry requires significant gas monitoring on the production floor, in research and development laboratories, and in plant utility rooms, especially because of its use of inert or toxic chemicals in often confined spaces. Detecting oxygen depletion is particularly important due to the use of nitrogen in many of these areas. Offering a range of gas detection products and configurations, VWR is well equipped to address the various hazards found in pharmaceutical production.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"24"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  Caron Products
Description:   Allows 2 tanks of CO2 or N2 to be connected. Automatically switches from primary tank to secondary tank when low gas pressure is detected, and sounds an alarm. Returns to primary tank upon detection of gas pressure at primary inlet. 115V, 60 Hz.
Small Business Enterprise
Supplier:  Hardy Diagnostics
Description:   Simple to use system for the rapid generation of an anaerobic environment necessary to cultivate anaerobic microorganisms.
Product available on GSA Advantage®

Supplier:  HARDY DIAGNOSTICS CA
Description:   The system consists of a plastic pouch and a paper gas generating sachet.
Catalog Number: (CA10790-378)

Supplier:  HARDY DIAGNOSTICS CA
Description:   Simple-to-use systems for the rapid generation of a microaerophilic environment necessary to cultivate Campylobacter species.
Supplier:  Sensidyne
Description:   Convenient, economical system tests the quality of compressed breathing air
Small Business Enterprise
Catalog Number: (CA56614-743)

Supplier:  3M
Description:   These personal monitoring devices contain a single charcoal-sorbent wafer for easy collection of organic vapors and analysis by gas chromatography (NI...
Supplier:  Draeger
Description:   For use with the accuro® Gas Detector Pump (see 34330-068 series) and Aerotest Breathing Air Tester (see 34330-078).
Supplier:  Draeger
Description:   Short-Term detector tubes for use with the accuro® Gas Detector Pump and Aerotest Breathing Air Tester.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 24
  1  2  3  Next 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping