Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • GSA Contract (55) UL Listed (30) C-UL Listed (25) CSA Certified (16) CE Compliant (9)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Sale Items
  • Search Within Results

Hotplates

https://ca.vwr.com/stibo/smallweb/std.lang.all/94/06/18989406.jpg

Less hazardous than baths, the hotplates uniformly heat substances for any required procedure. White ceramic surface options permit the slow titrant addition for concentration determinations and have the resistance to work with even corrosive chemicals. With superior durability to withstand accidental impacts, aluminum surface heating plates also spread high temperatures evenly. The economical hotplates are equipped with innovative safety features for efficient operation and workplace safety.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"36"
  List View Easy View
Sort by:
 
 
 
 

Catalog Number: (12620-980)

Supplier:  VWR International
Description:   The economical Dylatherm™ Hot Plate features a large 16.5 cm (6.5") diameter cast aluminum top plate that provides an even heating surface capable of ...
Product available on GSA Advantage® UL Listed CE Compliant
Catalog Number: (12620-978)

Supplier:  VWR International
Description:   The economical Dylatherm™ hot plate features a large 16.5 cm (61/2”) diameter cast aluminum top plate that provides an even heat...
Product available on GSA Advantage® UL Listed cUL Listed
Supplier:  VWR International
Description:   Advanced hot plates are designed to deliver accurate and repeatable results in general-purpose laboratory applications.
Product available on GSA Advantage®
Supplier:  VWR International
Description:   Rugged, compact hotplates with an ergonomic design are ideal for educational labs and can heat up to 1000 mL of water.
Product available on GSA Advantage®
Supplier:  VWR International
Description:   Designed for applications requiring exceptional accuracy, stability, and repeatability, these hot plates are equipped with an exclusive safety system ...
Product available on GSA Advantage®
Supplier:  VWR International
Description:   Standard hotplates are designed for general-purpose laboratory use.
Product available on GSA Advantage®
Supplier:  VWR International
Description:   Standard hotplates are designed for general-purpose laboratory use.
Product available on GSA Advantage® CE Compliant
Supplier:  VWR International
Description:   Advanced hotplates are designed to deliver accurate and repeatable results in general-purpose laboratory applications.
Product available on GSA Advantage® CE Compliant
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 36
  1  2  3  4  5  Next 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping