Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • Environmentally Preferable (33) GSA Contract (26) SBE (21) UL Listed (37) C-UL Listed (32) CSA Certified (14) Irregular Voltage (5)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Cooled Incubators

https://ca.vwr.com/stibo/smallweb/std.lang.all/50/70/18945070.jpg

Reaching as far down as freezing point, the cooled incubators create and control precise temperature conditions for the ideal refrigerated storage. These incubators can be used for long-term storage of drug substances, ultra-sensitive material, and temperature conditioning. Exterior digital displays are easy to read and manage. Glass door models allow personnel to constantly monitor contents. The cooled incubators come in a variety of sizes and volumes to fit any work space.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"17"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalog Number: (89511-416)
Supplier:  VWR International
Description:   Save space with a VWR® personal low temperature incubator ideal for small sample work.
Product available on GSA Advantage® CSA Certified
Supplier:  VWR International
Description:   The VWR® Signature™ Diurnal Growth Incubator is designed for studies requiring day and nighttime simulation.
Product available on GSA Advantage® Small Business Enterprise
Supplier:  VWR International
Description:   Using innovative peltier cooling technology, the VWR drosphila incubators offer a 78% reduction in energy consumption over traditional compressor base...
Small Business Enterprise
Supplier:  Caron Products
Description:   Caron’s Refrigerated Incubators precisely and tightly control temperature conditions from 5°C to 70°C with optional operation down to 0°C, making the...
Product is energy and/or water efficient Product available on GSA Advantage® Small Business Enterprise CSA Certified
Supplier:  Caron Products
Description:   Caron's extra large capacity Refrigerated Incubators precisely control temperature conditions
Product available on GSA Advantage® Small Business Enterprise
Supplier:  Binder
Description:   The KB refrigerated incubators for microorganisms are very versatile and offers a wide range of capabilities delivering reproducible test results
Product available on GSA Advantage® UL Listed
Supplier:  Binder
Description:   The KT refrigerated incubators with Peltier technology combine outstanding performance with environmental friendliness and provide sample protection.
Supplier:  Panasonic Healthcare
Description:   The Panasonic MIR series offers accurate temperature control and uniformity over a wide temperature range, making it suitable for various applications...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 17
  1  2  3  Next

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping