Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Microplate Incubators

https://ca.vwr.com/stibo/smallweb/std.lang.all/95/40/18989540.jpg

The microplate incubator is a cost-effective way to control environmental conditions for small sample sizes contained in Petri dishes and test tubes. Temperatures can easily be adjusted to meet sensitive material requirements. Interior heating elements provide the uniform, elevated temperatures for consistent batches. Microplate incubators are made of durable materials for heavy routine use and occupy a small surface area to fit even the spaces with minimum room.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"2"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalog Number: (12621-182)
Supplier:  VWR International
Description:   When placed inside a standard or CO2 incubator, this stability chamber works to reduce the variations often experienced in cell-based assay...
Product available on GSA Advantage®
Catalog Number: (35915-118)
Supplier:  Boekel
Description:   Ideal for small volume incubations at or near 37°C (98.6°F)
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 2  of 2
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping