Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Sale Items
  • Search Within Results
Inverted Microscopes
https://ca.vwr.com/stibo/smallweb/std.lang.all/69/17/20236917.jpg
Inverted microscopes are useful for observing living cells or organisms located at the bottom of a large container. This allows samples to remain in conditions that are more natural than the glass slides used with conventional upright alternatives. While the light source and condenser are located above the stage, the objectives are below and point upward. Providing a closer inspection of specimen, inverted microscopes come in a variety of eye and nose piece combinations.
SearchPresentationType-VERTICAL
SearchResultCount:"7"
  List View Easy View
Sort by:
Supplier:  VWR International
Description:   The VWR® Inverted Microscope is the ideal device for live cell and advanced applications, as well as routine laboratory, research, and educational nee...
Supplier:  VWR International
Description:   The VWR® Inverted Fluorescence Microscope is ideal for applications in biology, cell cytology, genetics, oncology, and immunology, and it is also well...
UL Listed cUL Listed CSA Certified
Supplier:  DISCOVER ECHO INC.
Description:   Revolve is the world’s first and only HYBRID microscope
Product available on GSA Advantage®
Supplier:  DISCOVER ECHO INC.
Description:   Revolve is the world’s first and only HYBRID microscope
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient Product available on GSA Advantage®
Supplier:  Motic
Description:   The Motic AE2000 Inverted microscope is the ideal instrument for routine live cell inspection in both educational and high-grade professional applicat...
UL Listed cUL Listed CSA Certified
Supplier:  Motic
Description:   With a wide range of potential applications, Motic’s AE31 Elite microscope is at home in both research and routine environments.
Catalog Number: (470007-822)
Supplier:  AMERICAN EDUCATIONAL PROD
Description:   Discover How to Affect the Images Received by the Eye
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address
VWR will not lift restrictions for residential shipping addresses.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 7  of 7