Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "Ventilation Filters"

https://ca.vwr.com/stibo/smallweb/std.lang.all/99/88/18989988.jpg

Ventilation filters are composed of fibrous materials designed to remove solid and chemical particulates from the air, removing bacteria, dust, and volatile organic compounds. Applications include general ventilation, pre-filters, stand-alone single-stage filtration, built-up frame banks, side access systems, rooftop air handlers, and air supply for laboratories. Various designs are available for meeting a variety of needs, such as applications where increased air velocity is present and low pressure drops are required. Highly durable and efficient.

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"4"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Labconco
Description:   Kit provides a second level of HEPA Filtration to meet United States Pharmacopeia (USP) <800> standard for redundant HEPA Filter in series. It...
Supplier:  Total Filtration Services
Description:   Activator™ series panels and links consist of layers of multi-denier polyester media sealed around a heavy-duty 9-gauge internal wire frame with integ...
Supplier:  Total Filtration Services
Description:   Extended surface pocket filters feature efficiency, high dust holding capacity, long service life, and low resistance to air flow.
Supplier:  Total Filtration Services
Description:   DP® filters are made of 100% synthetic fibers that provide mechanical efficiency for a MERV 8 performance.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 4  of 4