Would you like to log in again?
Nalgene® CryoBox™ Rack, Horizontal, Thermo Scientific
Supplier: NALGE NUNC INTERNATIONAL
Nalgene® CryoBox™
5038-4422 5038-4322
Stainless steel rack fits most upright freezer compartments
Warning: For safe storage in the liquid phase of liquid nitrogen, CryoTube® vials must be correctly sealed in CryoFlex™ tubing.
Handles at both ends for freezers with right- or left-hand doors. Holds standard 1–2mL cryoboxes. Shelves are arrayed in a 4x4 format.
ORDER
Expand All / Collapse All
W x D x H No. of Shelves per Rack Supplier No. VWR Catalog Number Unit Price Quantity
14.3 x 44.4 x 22.5 cm ( 55/8 x 171/2 x 87/8") 12 5038-4322 55710-170 Case of 2 Retrieving
Each Retrieving
14.3 x 59 x 22.5 cm ( 55/8 x 231/4 x 87/8")  16  5038-4422 55710-171 Case of 2 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty