Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Sale Items
  • Search Within Results

Distillation Receivers

https://ca.vwr.com/stibo/smallweb/std.lang.all/51/43/18945143.jpg

The distilling receivers are the containers that collect liquefied vapor in distillation apparatuses. Depending on the volume of organic liquid being purified, the glassware is available in single or multiple receiving formats to trap the moisture as it drips from the connected condenser. Tube styles allow access with syringes to recover substance, while flask style adapters are simply rotated to continue filling with an empty vessel. Distilling receivers come in standard joint sizes to fit other components incorporated.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"72"
  List View Easy View
Sort by:
 
 
 
 


Supplier:  Glassco
Description:   Distribution adapter, 24/40, Cow-type design features three receivers spaced 40° apart.
Supplier:  Glassco
Description:   Distilling Receivers have a 135° angle between the center line of the condenser and the receiver tubes
Catalog Number: (89426-360)

Supplier:  Glassco
Description:   Suitable for microscale application
Supplier:  Ace Glass
Description:   Arsenic Limit Test apparatus as described in the European Pharmacopeia chapter 2.4.3.
Small Business Enterprise
Supplier:  Ace Glass
Description:   This apparatus is used for a limit test for arsenic, as described in European Pharmacopeia, 1997, page 51
Product available on GSA Advantage® Small Business Enterprise
Supplier:  Wilmad-LabGlass
Description:   Made of borosilicate glass, these Bidwell-Sterling moisture traps are calibrated "to contain" (TC) at 20°C and are used for determining moisture in sa...
Catalog Number: (KT585151-0035)

Supplier:  DWK Life Sciences (KIMBLE®)
Description:   This distilling receiver is designed for Dean-Stark distillation.
Supplier:  Ace Glass
Description:   Especially useful when heavier-than-water solvents are employed
Product available on GSA Advantage® Small Business Enterprise
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 72
  1  2  3  4  5  6  7  8  9  Next 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping