Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Microscope Bulbs

Microscope bulbs options include tungsten incandescent, fluorescent, LED, and halogen styles to fit any existing equipment. These replacement bulbs will deliver clear lighting for detailed, up-close examinations of specimen. All models provide pinpoint lighting and adjustable light output to match any specific application. Light outputs are brilliant, cool, and perfectly balanced. The microscope bulbs insert easily into the optical instruments and have strong working longevities.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"5"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Bulbtronics
Description:   NARVA Model 55168 is a 30W, 6V, T3 Halogen bulb with a PY16-1.25 base
Supplier:  GE
Description:   Replacement bulbs for the microscopes with built-in illuminators.
Catalog Number: (470145-640)
Supplier:  Skywill Far East Industries
Description:   Additional Resources: -Grab-n-Go Assembly Instructions -For more information on purchasing the full kit, please visit the McGraw Hill website. ...
Catalog Number: (470021-696)
Supplier:  Walter Products
Description:   REPL BULB 20W TUNGSTEN FOR WARDS ADV
Catalog Number: (470021-692)
Supplier:  Walter Products
Description:   Inexpensive upgrade for mirror-illuminated microscope.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 5  of 5
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping