Cobalt ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), thickness 3.18 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

40834-KS 40834-KS
CAAA40834-KSEA 578.31 CAD
CAAA40834-KS AA40834-KS
Cobalt ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), thickness 3.18 mm (0.125 in)
Cobalt

Magnetron sputtering source

Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870 °C (1013 hPa)
Melting Pt: 1495 °C
Density: 8,9 g/cm³ (20 °C)
MDL Number: MFCD00010935
CAS Number: 7440-48-4
Merck Index: 12,02488

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR