Order Entry
ContactUsLinkComponent
Gallium Standard for ICP, Certipur®, MilliporeSigma
Gallium Standard for ICP, Certipur®, MilliporeSigma
Catalog #: CA1.70319.0100
Supplier:  MilliporeSigma
Gallium Standard for ICP, Certipur®, MilliporeSigma
Catalog #: CA1.70319.0100
Supplier:  MilliporeSigma
Price
$178.08
Log in for availability
Unit of Measure Each (100ml)
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    1000 µg/ml
  • Element:
    Gallium
  • Matrix:
    HNO₃ 2 - 3%
  • Packaging:
    Plastic Bottle
  • Size:
    100 ml
  • Cat. no.:
    CA1.70319.0100

Specifications

About this item

Traceable to SRM from NIST.

Certifications: The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
Ordering: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.
Customers Who Bought This Also Bought