Acrodisc® Units with Mustang® E Membrane, Cytiva (Formerly Pall Lab)

Supplier: Cytiva
Acrodisc®
MSTG25E3 MSTG25E3
CA28140-521PK 551.76 CAD
CA28140-521 28140-521
Acrodisc® Units with Mustang® E Membrane, Cytiva (Formerly Pall Lab)
Filters Syringe Filters
Disposible, single-use units remove endotoxins in water, buffers, neutral sugars, and certain biological solutions.

  • Disposable
  • Single-use
  • Gamma irradiated

Flow rate of 1 to 4 ml/min. (varying with type of solution, concentration of proteins, and other components).

Bed volume: 0.12 ml

Differential pressure: <0.07 bar (7 kPa, 1 psi) @ 1 ml/min.

Max. operating pressure: 2.1 bar (210 kPa, 30 psi) at 60 ˚C (140 ˚F) or 5.5 bar (550 kPa, 80 psi) at 21 to 24 ˚C (70 to 75˚F).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR