Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Acetophenone

SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
 
 
SearchResultCount:"4"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  MilliporeSigma
Description:   Acetophenone, CAS number:98-86-2, Synonyms:Methyl phenyl ketone, 1-Phenylethanone, Phenylethanone, ACP, Application:for synthesis
SDS
Catalog Number: (CA8.00028.9026)
Supplier:  MilliporeSigma
Description:   Acetophenone, CAS number:98-86-2, Synonyms:Methyl phenyl ketone, 1-Phenylethanone, Phenylethanone, ACP, Application:for synthesis
SDS
Supplier:  Alfa Aesar
Description:   ACETOPHENONE, 99% 500ML
Supplier:  TCI America
Description:   CAS Number: 98-86-2
MDL Number: MFCD00008724
Molecular Formula: C8H8O
Molecular Weight: 120.15
Purity/Analysis Method: >98...
SDS
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 4  of 4