Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Explorer® Precision Balances, Ohaus®

Supplier: OHAUS SCALE CORP
Explorer®
83021365 83021342 83021341 83021362 83021361 83021350 83021360 83021348 83021357 83021356
Precision balances provide accurate results in seconds to improve efficiency and productivity

Four touchless sensors allow for hands-free operation of print, tare, calibration, and other functions. All balances are equipped with AutoCal™ automatic internal calibration adjustment system, which self-calibrates the system when it senses a temperature change significant enough to affect weighing accuracy or every 11 hours. SmarText™ 2.0 graphical software features 14 applications, QWERTY and numeric keypads, and below-minimum weight indication. Large color touch screen displays allow for easy menu navigation. The display includes USB and RS-232 ports, and can be separated from the weighing base for maximum flexibility in a broad range of applications. The weighing base features a QuadraStance™ design with four adjustable thumbwheels to provide superior stability. All models offering a readability of 0.001g feature a frameless, flip-top draftshield constructed of antistatic coated glass

Certifications: CE marked. FCC, CMC, OIML Class I and Class II Approved.

Ordering information: Balances are supplied with two-year manufacturer's warranty, AC adapter, weighing pan, pan support, multilanguage operator's manual with quick-start guide, and in-use cover.



 

ORDER

Expand All / Collapse All
Weighing Capacity Linearity Pan/Platform Size Readability Stabilization Time Supplier No. VWR Catalog Number Unit Price Quantity
220 g ±0.002 g 13 cm (51/8") dia. 0.001 g 1.5 s 83021341 89171-452 Each Retrieving
420 g ±0.002 g 13 cm (51/8") dia. 0.001 g 1.5 s 83021342 89171-454 Each Retrieving
620 g ±0.002 g 13 cm (51/8") dia. 0.001 g 1.5 s 83021348 89171-456 Each Retrieving
1100 g ±0.002 g 13 cm (51/8") dia. 0.001 g 1.5 s 83021350 89171-458 Each Retrieving
2000 g ±0.02 g 19 x 20 cm (71/2 x 77/8") 0.01 g 1 s 83021356 89171-460 Each Retrieving
4200 g ±0.02 g 19 x 20 cm (71/2 x 77/8") 0.01 g 1 s 83021357 89171-462 Each Retrieving
6200 g ±0.1 g 19 x 20 cm (71/2 x 77/8") 0.1 g 1 s 83021361 89171-468 Each Retrieving
6200 g ±0.02 g 19 x 20 cm (71/2 x 77/8") 0.01 g 1 s 83021360 89171-464 Each Retrieving
10,200 g ±0.1 g 19 x 20 cm (71/2 x 77/8") 0.1 g 1 s 83021365 89171-470 Each Retrieving
10,200 g ±0.02 g 19 x 20 cm (71/2 x 77/8") 0.01 g 1 s 83021362 89171-466 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Dimensions 23 x 35 x 39.3 cm (91/16 x 13¾ x 15½") 23 x 9.8 x 39.3 cm (91/16 x 1327/32 x 15½") 23 x 35 x 39.3 cm (91/16 x 13¾ x 15½") 23 x 9.8 x 39.3 cm (91/16 x 1327/32 x 15½") 23 x 35 x 39.3 cm (91/16 x 13¾ x 15½")
Voltage 100–240V, 50–60Hz

ACCESSORIES

Expand All / Collapse All

Accessories for Explorer® Balances, Ohaus
Description Fits Supplier No. VWR Catalog Number Unit Price Quantity
Anti-Theft Device (Laptop lock), for Explorer®/Explorer® Pro/Triple Beam/MB Series All Explorer®/Explorer® Pro/Triple Beam/MB Series Models 80850043 11380-322 Each Retrieving
Cable, USB Interface, (Type A to B), for Explorer®/Adventurer® Series All Explorer®/Adventurer® Series Models 83021085 89194-600 Each Retrieving
Density Kit, Solids All Explorer® Models 80253384 89194-596 Each Retrieving
Ethernet Interface Kit, for Explorer® Series All Explorer® Models 83021082 89194-604 Each Retrieving
Ink Ribbon Casette for Printer SF40A, 57.5 mm Width, for Explorer®/Explorer® Pro/Discovery/Adventurer™ Pro/Pioneer™/Defender™/CKW/Ranger™/Valor™/Navigator®/Scout® Pro/MB Series All Explorer®/Explorer® Pro/Discovery/Adventurer™ Pro/Pioneer™/Defender™/CKW/Ranger™/Valor™/Navigator®/Scout® Pro/MB Series Models 12120798 89497-006 Each Retrieving
Paper Roll for Printer SF40A, 57.5 mm Width, for Explorer®/Explorer® Pro/Discovery/Adventurer™ Pro/Pioneer™/Defender™/CKW/Ranger™/Valor™/Navigator®/Scout® Pro/MB Series All Explorer®/Explorer® Pro/Discovery/Adventurer™ Pro/Pioneer™/Defender™/CKW/Ranger™/Valor™/Navigator®/Scout® Pro/MB Series Models 12120799 89497-004 Each Retrieving
Printer Cable for TXW, CKW All Explorer® Models 30041470 89496-998 Each Retrieving
Printer cable SF40A, for Discovery/Explorer®/Adventurer™ Pro/Pioneer™/Defender™ 3000/5000/MB series/Ranger™ Series All Discovery/Explorer®/Adventurer™ Pro/Pioneer™/Defender™ 3000/5000/MB series/Ranger™ Series Models 12122603 89497-008 Each Retrieving
Printer Replacement Cable All Explorer® Models 80850000 11380-304 Each Retrieving
Printer Replacement Ink Ribbon All Explorer® Models 30078078 89496-992 Each Retrieving
Rechargeable Battery, EX-HiCap R71 All Explorer® Models 30041295 89496-996 Each Retrieving
RS232 Cable, PC 25 Pin, for Discovery/Explorer®/Adventurer™ Pro/Pioneer™ Series All Discovery/Explorer®/Adventurer™ Pro/Pioneer™ Series Models 80500524 89194-608 Each Retrieving
RS232 Cable & Adapter to STP103 Printer, for Discovery/Explorer®/Adventurer™ Pro/Pioneer™/Defender™ 3000/5000/MB Series All Discovery/Explorer®/Adventurer™ Pro/Pioneer™/Defender™ 3000/5000/MB Series Models 80252580 97009-696 Each Retrieving
Second R232 Interface Kit, for Explorer® Series All Explorer® Models 83021081 89194-602 Each Retrieving
Terminal Extension Cable, for Explorer® Series All Explorer® Models 83021083 89194-594 Each Retrieving
Tower Mount for Terminal, for Explorer® Series All Explorer® Models 83021102 89194-592 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty