Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4,6-Trimethylbenzyl chloride, 98%

Supplier: ALFA AESAR CA
Synonyms: 1-(Chloromethyl)-2,4,6-trimethylbenzeneα-Chloro-1,2,4,6-tetramethylbenzene

L00423-06 L00423-14
Purity: 98%
CAS No.: 1585-16-6
Molecular Formula: C10H13Cl
Formula Weight: 168.67
Storage Temperature: 2°C to 8°C
Flash Point: 106°(222°F)
Odor: Pungent
Melting Point: 37-42°
Boiling Point: 114-115°/10mm
Beilstein Reference: 386678
UN No.: 3261
MDL No.: MFCD00000901


Formula: C₁₀H₁₃Cl
MW: 168.67 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 34 - 38 °C
Flash Pt: 106 °C (222 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000901
CAS Number: 1585-16-6
UN: 3261
ADR: 8,II


 

ORDER

Expand All / Collapse All
Size Packaging Supplier No. VWR Catalog Number Unit Price Quantity
25 g bottle L00423-14 CAAAAL00423-14 Each (25g) Retrieving Restricted
restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty