Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Electronic Balance Calibration Weights, ANSI/ASTM Class 2 Weight, Troemner

Supplier: TROEMNER LLC
8253 8254 8210 8216 8214 8252W 8240W 8252 8253T 8253W 8206 8224 8244W 8200W 8252T 8232W 8240T 8224T 8216W 8244T 8200T 8236W 8224W 8232T 8228T 8216T 8236T 8228W 8234 8232 8238 8236 8228 8200 8244 8242 8202 8240 8234W 8242T 8254W 8210W 8242W 8202T 8238W 8234T 8214W 8202W 8254T 8210T 8206T 8238T 8214T 8206W
Precision-crafted of stainless steel, with a highly polished finish

Weights 5kg and below supplied in a rugged, precision-fit polypropylene case with handling glove.

Standard weights are supplied with a Statement of Accuracy. Where applicable, weights are supplied with a NIST/NVLAP Weight Calibration Certificate or a Traceable Certificate.

Weights 30kg to 5kg are flat on top with a recessed grip handle for easy handling and stacking. Weights 5kg to 1kg are cylindrical in shape with a groove near the top to allow for easy handling and gripping. Weights below 1kg are straight cylinders. All are manufactured with adjusting cavities.



 

ORDER

Expand All / Collapse All
Certification Weight Supplier No. VWR Catalog Number Unit Price Quantity
NVLAP 160 g cyl 8240W 14216-614 Each Retrieving
NVLAP 100 g cyl 8244W 14216-590 Each Retrieving
NVLAP 30 kg grip 8200W 14216-740 Each Retrieving
NVLAP 5 kg cyl 8216W 14216-686 Each Retrieving
NVLAP 30 g cyl 8254W 14216-554 Each Retrieving
NVLAP 2 kg cyl 8224W 14216-674 Each Retrieving
NVLAP 25 kg grip 8202W 14216-728 Each Retrieving
NVLAP 400 g cyl 8234W 14216-650 Each Retrieving
NVLAP 500 g cyl 8232W 14216-662 Each Retrieving
NVLAP 5 kg grip 8214W 14216-692 Each Retrieving
NVLAP 50 g cyl 8252W 14216-578 Each Retrieving
NVLAP 40 g cyl 8253W 14216-566 Each Retrieving
NVLAP 10 kg grip 8210W 14216-704 Each Retrieving
NVLAP 300 g cyl 8236W 14216-638 Each Retrieving
NVLAP 150 g cyl 8242W 14216-602 Each Retrieving
NVLAP 200 g cyl 8238W 14216-626 Each Retrieving
NVLAP 1000 g cyl 8228W 14216-668 Each Retrieving
NVLAP 20 kg grip 8206W 14216-716 Each Retrieving
Statement of Accuracy 2 kg cyl 8224 14216-670 Each Retrieving
Statement of Accuracy 25 kg grip 8202 14216-724 Each Retrieving
Statement of Accuracy 160 g cyl 8240 14216-610 Each Retrieving
Statement of Accuracy 5 kg grip 8214 14216-688 Each Retrieving
Statement of Accuracy 30 g cyl 8254 14216-550 Each Retrieving
Statement of Accuracy 200 g cyl 8238 14216-622 Each Retrieving
Statement of Accuracy 400 g cyl 8234 14216-646 Each Retrieving
Statement of Accuracy 50 g cyl 8252 14216-574 Each Retrieving
Statement of Accuracy 300 g cyl 8236 14216-634 Each Retrieving
Statement of Accuracy 150 g cyl 8242 14216-598 Each Retrieving
Statement of Accuracy 1000 g cyl 8228 14216-664 Each Retrieving
Statement of Accuracy 5 kg cyl 8216 14216-682 Each Retrieving
Statement of Accuracy 30 kg grip 8200 14216-736 Each Retrieving
Statement of Accuracy 10 kg grip 8210 14216-700 Each Retrieving
Statement of Accuracy 40 g cyl 8253 14216-562 Each Retrieving
Statement of Accuracy 500 g cyl 8232 14216-658 Each Retrieving
Statement of Accuracy 20 kg grip 8206 14216-712 Each Retrieving
Statement of Accuracy 100 g cyl 8244 14216-586 Each Retrieving
Traceable 100 g cyl 8244T 14216-588 Each Retrieving
Traceable 200 g cyl 8238T 14216-624 Each Retrieving
Traceable 2 kg cyl 8224T 14216-672 Each Retrieving
Traceable 5 kg cyl 8216T 14216-684 Each Retrieving
Traceable 300 g cyl 8236T 14216-636 Each Retrieving
Traceable 20 kg grip 8206T 14216-714 Each Retrieving
Traceable 30 g cyl 8254T 14216-552 Each Retrieving
Traceable 40 g cyl 8253T 14216-564 Each Retrieving
Traceable 400 g cyl 8234T 14216-648 Each Retrieving
Traceable 25 kg grip 8202T 14216-726 Each Retrieving
Traceable 5 kg grip 8214T 14216-690 Each Retrieving
Traceable 10 kg grip 8210T 14216-702 Each Retrieving
Traceable 500 g cyl 8232T 14216-660 Each Retrieving
Traceable 150 g cyl 8242T 14216-600 Each Retrieving
Traceable 30 kg grip 8200T 14216-738 Each Retrieving
Traceable 160 g cyl 8240T 14216-612 Each Retrieving
Traceable 50 g cyl 8252T 14216-576 Each Retrieving
Traceable 1000 g cyl 8228T 14216-666 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
Optional Case for 8 and 10 kg TW-50-10KG 12765-500 Each Retrieving
Optional Case for 16 and 20 kg TW-100-20KG 12765-502 Each Retrieving
Optional Case for 25 kg TW-100-25KG 12765-504 Each Retrieving
Optional Case for 30 kg TW-100-30KG 12765-506 Each Retrieving
Replacement Case for 600 - 10 g Individual Weights 8C1000 12985-000 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SERVICES


Services: VWR Catalyst Services provides convenient and fast re-calibration services within 6 business days for your weights. Please contact us at 1.888.793.2300 or VWRCATALYST™@vwr.com for a quote or more information. If your weights are found to be out of tolerance, we will make the proper adjustments to bring them back into tolerance. To prevent loss and ensure the timely processing and integrity of your precision weights and/or calibration devices, please do not pack in foam peanuts. Please avoid packing your items with foam, packing peanuts or other material that cannot be recycled.

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty