Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HC-75 Hydrogen Form Cation Exchange HPLC Columns, Hamilton Company

Supplier: HAMILTON COMPANY
79476 79544 79547
Hamilton Company offers one of the most comprehensive selections of chromatography columns in the industry.

  • Separate Mono and Disaccharides
  • Separate oligomers from DP1 to DP8
  • Compatible with RI detectors
  • 9 µm particle size
  • Multiple column internal diameters available
  • Stainless Steel or PEEK hardware available
  • Buffer strength 0.0 - 0.05N

Each column is manufactured under rigorous criteria and tested for resolution and retention time accuracy.

This product employs polymeric cross-linked soft-gel materials for cation, ligand exchange separation of carbohydrates.

HC-75 gel-type cation exchange columns are especially useful for determining ethanol in fermentation broths and for separating sugars and sugar alcohols in food samples like com syrup, chewing gum sweeteners, and fruit juices.

HC-75 columns separate compounds through size exclusion and ligand exchange. Ligand exchange dominates in the more highly cross-linked HC-75.




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Description Column Format Column Hardware Phase I.D. x L Packing Material Particle Size Bed Weight Supplier No. VWR Catalog Number Unit Price Quantity
Hamilton HC-75 Hydrogen Form Cation Exchange HPLC Column; 100% Water 0-40% Acetonitrile Analytical Stainless steel PSDVB/Sulfonate Exchanger 7.8 x 100 mm PSDVB/Sulfonic Acid 9 µm 5 meq/gm 79547 21492-536 Each Retrieving
Hamilton HC-75 Hydrogen Form Cation Exchange HPLC Column; 100% Water 0-40% Acetonitrile Analytical Stainless steel PSDVB/Sulfonate Exchanger 7.8 x 305 mm PSDVB/Sulfonic Acid 9 µm 5 meq/gm 79544 89221-154 Each Retrieving
Hamilton HC-75 Hydrogen Form Cation Exchange HPLC Column; 100% Water 0-40% Acetonitrile Analytical Stainless steel PSDVB/Sulfonate Exchanger 4.1 x 250 mm PSDVB/Sulfonic Acid 9 µm 5 meq/gm 79476 89221-136 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description 21492-536 89221-154 89221-136
Max. pressure 400 psi
Temperature limit 25-90°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty