Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nalgene® Diagnostic Bottles Natural HDPE, Without Closures Bulk Pack, Thermo Scientific

Supplier: NALGE NUNC INTERNATIONAL
362002-9125 362002-9025 362002-9050
Nalgene Diagnostic Bottles are used in test kits and many small-volume applications

Diagnostic bottles can be used for sample storage and shipping of small volume, valuable reagents. These bottles are packed without closures so they can be matched with various colored closures.

Ordering information: Bulk packed for packaging applications.



 

ORDER

Expand All / Collapse All
Capacity Height with Cap Neck I.D. Outer Diameter Cap Size Supplier No. VWR Catalog Number Unit Price Quantity
15 mL (0.5 oz.) 58 mm 14 mm 25 mm 20 mm 362002-9050 16211-004 Case of 2,000 Retrieving
4 mL (0.14 oz.) 41 mm 8 mm 16 mm 13 mm 362002-9125 16211-006 Case of 2,000 Retrieving
8 mL (0.27 oz.) 45 mm 14 mm 25 mm 20 mm 362002-9025 73521-136 Case of 2,000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping