Printer Kit for Advanced Microlab® 600 Dispenser/Diluter and Microlab® 300 Guided Pipetting System, Hamilton Company
Supplier: HAMILTON COMPANY
61500-04
89209-582EA 1472.26 CAD
89209-582
Printer Kit for Advanced Microlab® 600 Dispenser/Diluter and Microlab® 300 Guided Pipetting System, Hamilton Company
Printers
The printer kit works with the Advanced Microlab® 600 diluter/dispenser and with the Microlab 300 Guided Pipetting System.
Microlab® 600 diluter/dispenser printer kit interfaces with the free compliance software package that is available for Advanced devices. The kit automatically is compatible with all Microlab 300 pipettors.
The printer kit satisfies regulatory CFR21 Part 11 and FDA GMP/GLP requirements by conveniently printing all log files directly from the device's controller.
The printer prints on standard, non-thermal 8.5x11 and A4 paper.
Certifications: CE and CCC Certified
Ordering information: The printer kit includes a printer, power source, adapter and cable.