Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Handbooks and Binders

https://ca.vwr.com/stibo/smallweb/std.lang.all/93/84/18989384.jpg

Containing all the required information in one location, handbooks and binders allow personnel to instantly reference particular codes of behavior or step-by-step operating techniques. Binders store important documents in a protected method for guaranteed longevity even with heavy every day use. Using structured handbooks will establish expectations while protecting employers from possible liability issues. Available in common languages, the advantageous handbooks and binders successfully communicate standards to all personnel.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"9"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Brady Worldwide
Description:   Our standard MSDS binders are available in different sizes to suit all of your needs
Supplier:  American Compliance Systems
Description:   A comprehensive ergonomic program can help lower costs and increase productivity in the workplace.
Catalog Number: (CA82004-298)
Supplier:  Brady Worldwide
Description:   Our MSDS Training Binder Insert package contains OSHA Haz-Com standard highlights, compliance checklist, one 15-page RTK training booklet in English, ...
Supplier:  National Marker
Description:   Provide your employees with helpful information they need for optimum safety
Catalog Number: (CA89007-454)
Supplier:  Brady Worldwide
Description:   Use our Lockout/Tagout Handbook as a guide during training sessions to teach workers how to protect themselves from hazardous energy
Catalog Number: (CA82004-340)
Supplier:  Brady Worldwide
Description:   Made from heavy-duty bright red poly, this printed clipboard is easy to spot in an emergency situation.
Supplier:  National Marker
Description:   A central reference location for Material Safety Data Sheets
Supplier:  Brady Worldwide
Description:   The binder comes with a durable polyethylene cover, 36" chain and a pair of sheet lifters.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 9
  1  2  Next