Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • GSA Contract (8) MWBE (42) SBE (42)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Cell Culture Buffers

https://ca.vwr.com/stibo/smallweb/std.lang.all/99/17/18989917.jpg

Tissue culture buffers provide an energy source, maintain proper pH levels, and keep ideal osmotic pressure present. Used predominately in biomolecular research, the dry and liquid substances have a balanced inorganic salt composition. Lower concentration options permit equilibration with air in a closed system, while higher salt concentration options keep gaseous phases in equilibrium. Cell growth is optimal when plated in a nutritional medium that has been buffered.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"67"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   Blood bank saline is pH-controlled for a pH between 6.5 and 7.0.
SDS
Catalog Number: (BDH7895-4)
Supplier:  VWR International
Description:   Made with sodium phosphate monobasic monohydrate, sodium hydroxide solution and deionized water.
SDS
Supplier:  VWR
Description:   Hanks' Balanced Salt Solution (HBSS) is a buffer used to maintain a physiological pH for cells maintained in non-CO2 atmospheric conditions.
SDS
Product available on GSA Advantage®
Supplier:  VWR
Description:   DPBS is a balanced salt solution (BSS) used for the handling and culturing of mammalian cells.
SDS
Product available on GSA Advantage®
Supplier:  Ward's Science
Description:   Buffer solution has a shelf life of 24 months
SDS
Supplier:  Ward's Science
Description:   PHOSPHATE BUFFERED SALINE PH7.2 LG 500ML
SDS
Supplier:  VWR
Description:   1X Solution Composition:137 mM NaCl, 2.7 mM KCl, 9.5 mM Phosphate buffer.
Product available on GSA Advantage®
Supplier:  Ward's Science
Description:   Phosphate Buffer 0.1 M, Lab Grade 1 L^
SDS
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 67
  1  2  3  4  5  6  7  8  9  Next