Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "nucleic-acid-purification-reagents"

https://ca.vwr.com/stibo/smallweb/std.lang.all/95/96/18989596.jpg

Nucleic acids may be purified through a variety of different methods, and the reagents serve to make these processes more effective and efficient. The solutions amplify DNA or RNA strands and reduce the time and labor needed for template preparation. These methods often include the isolation of the nucleic acid in question prior to purification, with certain reagents being free of DNAse, RNAse, and protease altogether.

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"31"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Quantabio
Description:   PerfeCTa® DNase I is a recombinant bovine DNase I preparation that is free of any contaminating RNases or proteases.
Supplier:  Quantabio
Description:   PerfectPure RNA Cultured Cell Kit 50 prep, Kit content: Lysis Solution1, Wash 1 Solution, Wash 2 Solution (5 ml), Elution Solution, Purification Columns in Clear Tube, Collection Tubes, Lyophilized DNase, DNase Buffer Solution, DNase Wash Solution.
Limited Availability
Supplier:  Quantabio
Description:   PerfectPrep EndoFree Plasmid Maxi Kit 10 PREP, Kit content: RNase A, Buffer BL, Buffer PR1, Buffer PL2, Buffer PN4, Buffer PW (70 ml), Buffer PEB (30 ml), PerfectPrep EndoFree Filter CS, PerfectPrep Plasmid Mini Columns CB5, 50 ml Collection Tube.
SDS
Limited Availability
Supplier:  GE Healthcare - Life Sciences
Description:   Single Cell GenomiPhi DNA Amplification kit combines the unique capabilities of Phi29 DNA polymerase with an optimized formulation to amplify genomic ...
SDS
Supplier:  GE Healthcare - Life Sciences
Description:   RAPD reactions are pre-optimized for use with a wide variety of organisms. For performing DNA profiling experiments using the randomly amplified polym...
Supplier:  GE Healthcare - Life Sciences
Description:   (High-throughput) preparation of circular DNA templates for cycle sequencing, cloning, and transformation in 4-8 h depending on starting material, 16-...
Catalog Number: (CA95016-998L)
Supplier:  GE Healthcare - Life Sciences
Description:   illustra CsTFA is a strong chaotrope that not only inhibits ribonucleases, but also promotes the hydration and solubilization of nucleic acids and pro...
SDS
Supplier:  Omega Bio-Tek
Description:   BUFFER NL ^
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 31
  1  2  3  4  Next