  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Refractive Index Standards
https://ca.vwr.com/stibo/smallweb/std.lang.all/97/26/18989726.jpg
Prior to every analysis, use refractive index standards to check refractometer precision in evaluating sugar content. The known concentrations will verify a zero baseline reading so subsequent readings can be compared correctly for production consistency. While sucrose solutions evaluate devices that do not require a controlled temperature, calibration oils test instruments that involve specific levels. Refractive index standards guarantee that customers receive accurate Brix scale reports from the food industry.
SearchPresentationType-VERTICAL
SearchResultCount:"6"
  List View Grid View Easy View (new)
Sort by:
Supplier:  MilliporeSigma
Description:   Refractive index standard, traceable to SRM from NIST and PTB 50degree Brix Certipur*, Pack size: 1ML
Supplier:  MilliporeSigma
Description:   Certipur Refractive Index Standard Kit 2 Toluene/Water, Traceable to Srm From Nist and Ptb 1St^
SDS
Supplier:  MilliporeSigma
Description:   Certipur Refractive Index Standard Kit 3 1-Methyl-Naphthalene/Water, Traceable to Srm From Nist and Ptb
SDS
Supplier:  MilliporeSigma
Description:   Certipur Refractive Index Standard Kit 1 2, 2, 4-Trimethyl-Pentane/Water, Traceable to Srm From Nist and Ptb
SDS
Supplier:  Sper Scientific
Description:   Solution is packaged 10 bottles per box.
Catalog Number: (47735-584)
Supplier:  Cargille Laboratories
Description:   Refractive index liquids have a broad range of applications in diversified fields.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 6  of 6