Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Graphic See Signs, NMC

Supplier: NATIONAL MARKER COMP.
S92R S90P S91R S72R S92P S93R S91P S72P S74R S71P S93P S73R S95R S98R S74P S75R S73P S95P S9P S7P S90R S7R S98P S75P
Identify hazards, and areas of access for disable employees and visitors.



 

ORDER

Expand All / Collapse All
Material Dimensions Legend Supplier No. VWR Catalog Number Unit Price Quantity
PS Vinyl 17.78 x 17.78 cm (7 x 7") Biohazard No Food Or Drink To Be Stored.. (W/Graphic) S71P 94020-702 Each Retrieving
PS Vinyl 17.78 x 17.78 cm (7 x 7") No Exit (W/Graphic) S7P 94020-724 Each Retrieving
PS Vinyl 17.78 x 17.78 cm (7 x 7") Men (W/Graphic) S73P 94020-710 Each Retrieving
PS Vinyl 17.78 x 17.78 cm (7 x 7") Biohazard Authorized Personnel Only (W/Graphic) S93P 94020-742 Each Retrieving
PS Vinyl 17.78 x 17.78 cm (7 x 7") Hard Hat Required (W/Graphic) S9P 94020-754 Each Retrieving
PS Vinyl 17.78 x 17.78 cm (7 x 7") Volume Controlled (W/Graphic) S98P 94020-750 Each Retrieving
PS Vinyl 17.78 x 17.78 cm (7 x 7") Access Ramp (W/Graphic) S72P 94020-706 Each Retrieving
PS Vinyl 17.78 x 17.78 cm (7 x 7") Biohazard Peligro Biologico (Bilingual) (W/Graphic) S91P 94020-734 Each Retrieving
PS Vinyl 17.78 x 17.78 cm (7 x 7") Women (W/Graphic) S74P 94020-714 Each Retrieving
PS Vinyl 17.78 x 17.78 cm (7 x 7") Biohazard Use Universal Precautions (W/Graphic) S95P 94020-746 Each Retrieving
PS Vinyl 17.78 x 17.78 cm (7 x 7") Restrooms (W/Graphic) S75P 94020-718 Each Retrieving
PS Vinyl 17.78 x 17.78 cm (7 x 7") Biohazard Dispose Of Properly (W/Graphic) S92P 94020-738 Each Retrieving
PS Vinyl 17.78 x 17.78 cm (7 x 7") Biohazard Sharps (W/Graphic) S90P 94020-730 Each Retrieving
Rigid Plastic 17.78 x 17.78 cm (7 x 7") Biohazard Peligro Biologico (Bilingual) (W/ Graphic) S91R 94020-736 Each Retrieving
Rigid Plastic 17.78 x 17.78 cm (7 x 7") Biohazard Sharps (W/ Graphic) S90R 94020-732 Each Retrieving
Rigid Plastic 17.78 x 17.78 cm (7 x 7") Volume Controlled (W/ Graphic) S98R 94020-752 Each Retrieving
Rigid Plastic 17.78 x 17.78 cm (7 x 7") Biohazard Authorized Personnel Only (W/ Graphic) S93R 94020-744 Each Retrieving
Rigid Plastic 17.78 x 17.78 cm (7 x 7") Restrooms (W/ Graphic) S75R 94020-720 Each Retrieving
Rigid Plastic 17.78 x 17.78 cm (7 x 7") No Exit (W/ Graphic) S7R 94020-726 Each Retrieving
Rigid Plastic 17.78 x 17.78 cm (7 x 7") Men (W/ Graphic) S73R 94020-712 Each Retrieving
Rigid Plastic 17.78 x 17.78 cm (7 x 7") Biohazard Use Universal Precautions (W/ Graphic) S95R 94020-748 Each Retrieving
Rigid Plastic 17.78 x 17.78 cm (7 x 7") Access Ramp (W/ Graphic) S72R 94020-708 Each Retrieving
Rigid Plastic 17.78 x 17.78 cm (7 x 7") Biohazard Dispose Of Properly (W/ Graphic) S92R 94020-740 Each Retrieving
Rigid Plastic 17.78 x 17.78 cm (7 x 7") Women (W/ Graphic) S74R 94020-716 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty