Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • SBE (1)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

HPLC Sorbents

https://ca.vwr.com/stibo/smallweb/std.lang.all/53/12/18945312.jpg

By pumping pressurized liquid solvent with the sample mixture through a HPLC sorbent-filled column, researchers are able to identify each mixture component using retention time measurements. These sorbents are capable of working with small particle sizes requiring high operational pressure, a common high-performance liquid chromatography operation characteristic. Able to separate and quantify, the HPLC sorbents have a hydrophobic or polar nature. The versatile materials function with digital microprocessing instruments for rapid testing.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"22"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Avantor Performance Materials
Description:   Mixed Bed Exchange Resin, IONAC NM-60. BAKER ANALYZED* H+/OH- Form, Type I, Beads (16-50 mesh), Strong Acid/Strong Base; Sulfonated Alkyl Quaternary Ammoniom Polystyrene. 500g.
SDS
Supplier:  Avantor Performance Materials
Description:   Wide-Pore Butyl (C4) chromatographic media is made by covalently bonding butyl ligands to pure silica gel using a proprietary process to optimize sepa...
SDS
Supplier:  MilliporeSigma
Description:   LiChrospher® is the name given to reliable and versatile traditionally produced spherical silica carriers. LiChrospher® silica carriers are available ...
SDS
Supplier:  MilliporeSigma
Description:   Superspher® is a high performance silica carrier with a particle size of 4 µm which yields an excellent pressure/separation performance ratio.
Supplier:  MilliporeSigma
Description:   LiChrosorb® Si 100 is a reliable irregular shaped silica sorbent with a pore diameter of 100 Å for normal phase chromatography.
SDS
Supplier:  MilliporeSigma
Description:   EXtrelut® NT packing material is used for the production of large extraction columns.
Catalog Number: (CA95056-244L)
Supplier:  GE Healthcare - Life Sciences
Description:   NHS HP SpinTrap Buffer kit^
Supplier:  GE Healthcare - Life Sciences
Description:   Pre-made buffers optimized for the affinity-based enrichment of a target protein/immunoprecipitation using Protein A HP, Protein G HP, and Ab SpinTrap...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 22
  1  2  3  Next