Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Product Category
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "Antigen-Antibody Tests"

https://ca.vwr.com/stibo/smallweb/std.lang.all/49/01/18944901.jpg

Antigen-antibody tests rely on the fact that there is a specific antibody for each antigen, and so each one can be used to detect the presence of the other. These tests are usually performed on serum, but occasionally samples of other body fluids, tissue, or stool may be used. Different models of antigen-antibody tests are available for the isolation and differentiation of various strains of infectious diseases. Superior testing kits deliver efficient detection of multiple antigens.

SearchPresentationType-SearchPresentationType-HORIZONTAL
 
 
SearchResultCount:"25"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Hardy Diagnostics
Description:   Cefaclor CEC-30 HardyDisks* AST 1x50 disk
Catalog Number: (CA10693-108)
Supplier:  HARDY DIAGNOSTICS CA
Description:   Buffer No 6 500ml Poly carbonate Bottle
Catalog Number: (CA10693-106)
Supplier:  HARDY DIAGNOSTICS CA
Description:   Buffer No 1 500ml Poly carbonate Bottle
Catalog Number: (CA10693-110)
Supplier:  HARDY DIAGNOSTICS CA
Description:   Buffer No 16, USP (81), 500 ml Poly carbonate Bottle
Supplier:  HARDY DIAGNOSTICS CA
Description:   BCSA
Supplier:  Enzo Life Sciences
Description:   SAView* (mouse/rabbit-HRP, DAB) IHC kit, an immunohistochemistry product
Catalog Number: (CA11027-062)
Supplier:  BD
Description:   Tube Enteropluri Test Pk25
Supplier:  DIACARTA
Description:   QClamp* JAK2 Codon specific mutation test 10 samples
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 25
  1  2  3  4  Next