Would you like to log in again?
Cincinnati Form Franklin Flask
Supplier: STEINER ENTERPRISES SE
CP77775-00
Demonstrate boiling at a reduced temperature and pressure.
  • Requires support ring with clamp
This specially designed 1-liter flask is filled with water.
After the water is brought to a boil the neck is closed off with a rubber stopper fitted with a thermometer. Ice is then placed in the special concave bottom. As you do so, the water begins to boil rapidly again. The thermometer clearly shows that the temperature is falling but the water will boil until it is only 15-20° above room temperature.
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Cincinnati Form Franklin Flask CP77775-00 470007-718 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Size 12 cm
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty