Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Anatomy Activity Models

Supplier: AMERICAN EDUCATIONAL PROD
2677 2676 2675 2674 2630 2673 2640 2650 2672 2671
These raised-relief models are great for teaching basic anatomy or for classroom display.

  • Each Model Set Shows a Different Anatomical System
  • Models Sized for Easy Use in a Lab Group or Station
  • Teacher Materials and Suggested Student Activities Included
  • Raised Relief-Models
  • System or Specific-Organ Models Available

The model does not display the key, so it’s a great fit for practicals. System or specific-organ models are available.

Each kit includes a raised-relief model, an activity binder with lesson plans, copymasters for student activities, and overhead transparencies.

Ordering information: This kit includes a raised-relief model, an activity binder containing lesson plans, copy masters for student activities, and overhead transparencies.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Description Length Width No. of Parts Supplier No. VWR Catalog Number Unit Price Quantity
Circulatory System Activity 61 cm 46 cm 1 2671 470006-286 Each Retrieving
Respiratory System Activity 46 cm 61 cm 1 2672 470007-618 Each Retrieving
Digestive System Activity 46 cm 61 cm 1 2673 470007-620 Each Retrieving
Urinary System Activity 61 cm 46 cm 1 2675 470006-292 Each Retrieving
Nervous System Activity 61 cm 46 cm 1 2674 470002-706 Each Retrieving
Endocrine System Activity 46 cm 61 cm 1 2677 470002-704 Each Retrieving
Skin Activity 46 cm 61 cm 1 2630 470002-710 Each Retrieving
Ear Activity 46 cm 61 cm 1 2650 470007-616 Each Retrieving
Eye Activity 46 cm 61 cm 1 2640 470004-152 Each Retrieving
Teeth Activity 61 cm 46 cm 1 2676 470002-712 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty